CAS RN 18171-11-4
IUPAC Name:
(chloromethyl)dimethyl(propan-2-yloxy)silane
Synonyms:
(chloromethyl)(isopropoxy)dimethylsilane
Molecular Weight (g/mol):
166.72
Molecular Formula:
C6H15ClOSi
InChi Key:
KOTFCAKXPLOYLN-UHFFFAOYSA-N
SMILES:
CC(C)O[Si](C)(C)CCl
1
–
1
of
1
results
1
(Chloromethyl)isopropoxydimethylsilane 96.0+%, TCI America™
CAS: 18171-11-4 Molecular Formula: C6H15ClOSi Molecular Weight (g/mol): 166.72 MDL Number: MFCD00010605 InChI Key: KOTFCAKXPLOYLN-UHFFFAOYSA-N Synonym: (Chloromethyl)dimethylisopropoxysilane PubChem CID: 266137 IUPAC Name: chloromethyl-dimethyl-propan-2-yloxysilane SMILES: CC(C)O[Si](C)(C)CCl