Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethyl)isopropoxydimethylsilane 96.0+%, TCI America™
SDP

Catalog No. C13705ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13705ML 5 mL
C137025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13705ML Supplier TCI America Supplier No. C13705ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18171-11-4
Molecular Formula C6H15ClOSi
Molecular Weight (g/mol) 166.72
MDL Number MFCD00010605
InChI Key KOTFCAKXPLOYLN-UHFFFAOYSA-N
Synonym (Chloromethyl)dimethylisopropoxysilane
PubChem CID 266137
IUPAC Name chloromethyl-dimethyl-propan-2-yloxysilane
SMILES CC(C)O[Si](C)(C)CCl

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 mL
UN Number 1993
Formula Weight 166.72
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)isopropoxydimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.