accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CSiCH3CH3HO

CAS RN 18173-64-3

IUPAC Name: tert-butyldimethylsilanol
Synonyms: tert-butyldimethylsilanol
Molecular Weight (g/mol): 132.28
Molecular Formula: C6H16OSi
InChi Key: FGWRMMTYIZKYMA-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)