Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butyldimethylsilanol

Catalog No. 2890945G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-909-45G 5 g
28-909-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 28-909-45G Supplier Sigma Aldrich Supplier No. 2890945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyldimethylsilanol can be used as a silylating agent for the protection of hydroxyl groups via silylation. It is used:For the preparation of +a-chiral ether derivatives by catalytic asymmetric allylic substitution.As an initiator for the polymerization of 1,2 benzenedicarboxaldehyde.In the synthesis of enol silyl ethers.

Specifications

CAS 18173-64-3
Density 0.84 g/mL at 25°C
Boiling Point 139°C (739 mmHg, lit.)
Molecular Formula C6H16OSi
Linear Formula (CH3)3CSi(CH3)2OH
Refractive Index n20/D 1.424
MDL Number MFCD00010673
Quantity 5 g
Molecular Weight (g/mol) 132.28 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.