accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBrI

CAS RN 181765-86-6

IUPAC Name: methyl 5-bromo-2-iodobenzoate
Synonyms: methyl 5-bromo-2-iodobenzoate
Molecular Weight (g/mol): 340.94
Molecular Formula: C8H6BrIO2
InChi Key: CJRHLSZJEFJDLA-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(I)C=CC(Br)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results