Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Bromo-2-iodobenzoate 95.0+%, TCI America™
SDP

Catalog No. M30845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30845G 5 g
M30841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30845G Supplier TCI America Supplier No. M30845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 181765-86-6
Molecular Formula C8H6BrIO2
Molecular Weight (g/mol) 340.94
MDL Number MFCD00144771
InChI Key CJRHLSZJEFJDLA-UHFFFAOYSA-N
Synonym 5-Bromo-2-iodobenzoic Acid Methyl Ester
PubChem CID 11078356
IUPAC Name methyl 5-bromo-2-iodobenzoate
SMILES COC(=O)C1=C(I)C=CC(Br)=C1

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 340.94
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Bromo-2-iodobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.