accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFCH3NH2

CAS RN 1817801-55-0

IUPAC Name: 4-fluoro-2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 4-fluoro-2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 251.11
Molecular Formula: C13H19BFNO2
InChi Key: MONFQILBYDTFKM-UHFFFAOYSA-N
SMILES: CC1=C(N)C=C(B2OC(C)(C)C(C)(C)O2)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results