accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 181817-14-1

OOOOHOONH(S)(R)CH3CH3

CAS RN 181817-14-1

IUPAC Name: (2S,3R)-3-(acetyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2S,3R)-3-(acetyloxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 383.4
Molecular Formula: C21H21NO6
InChi Key: ZITLPPLNBYOLDJ-BLVKFPJESA-N
SMILES: C[C@@H](OC(C)=O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)