accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 181828-01-3

IUPAC Name: 4-(dimethylamino)-1-(2,2,2-trifluoroacetyl)pyridin-1-ium trifluoroacetate
Synonyms: 4-(dimethylamino)-1-(2,2,2-trifluoroacetyl)pyridin-1-ium trifluoroacetate
Molecular Weight (g/mol): 332.2
Molecular Formula: C11H10F6N2O3
InChi Key: GOSOPAPEXLJAEZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(F)(F)F.CN(C)C1=CC=[N+](C=C1)C(=O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results