Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trifluoroacetyl)-4-(dimethylamino)pyridinium Trifluoroacetate 98.0+%, TCI America™
SDP

Catalog No. T26025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26025G Supplier TCI America Supplier No. T26025G

Chemical Identifiers

CAS 181828-01-3
Molecular Formula C11H10F6N2O3
Molecular Weight (g/mol) 332.202
MDL Number MFCD01321272
InChI Key GOSOPAPEXLJAEZ-UHFFFAOYSA-M
PubChem CID 10860529
IUPAC Name 1-[4-(dimethylamino)pyridin-1-ium-1-yl]-2,2,2-trifluoroethanone;2,2,2-trifluoroacetate
SMILES CN(C)C1=CC=[N+](C=C1)C(=O)C(F)(F)F.C(=O)(C(F)(F)F)[O-]

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 5 g
Formula Weight 332.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Trifluoroacetyl)-4-(dimethylamino)pyridinium Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.