accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOHOCH3H3C

CAS RN 1820574-15-9

IUPAC Name: 2-[2,2-bis(propan-2-yl)-1,3-dioxolan-4-yl]ethan-1-ol
Synonyms: 2-(2,2-diisopropyl-1,3-dioxolan-4-yl)ethanol
Molecular Weight (g/mol): 202.29
Molecular Formula: C11H22O3
InChi Key: XFPZNMFFBDUEGH-UHFFFAOYNA-N
SMILES: CC(C)C1(OCC(CCO)O1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results