Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane 95.0+%, TCI America™
SDP

Catalog No. D40761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40761G Supplier TCI America Supplier No. D40761G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1820574-15-9
Molecular Formula C11H22O3
Molecular Weight (g/mol) 202.29
MDL Number MFCD22041646
InChI Key XFPZNMFFBDUEGH-UHFFFAOYNA-N
Synonym (R)-2-(2,2-Diisopropyl-1,3-dioxolan-4-yl)ethanol
PubChem CID 56973691
IUPAC Name 2-[2,2-bis(propan-2-yl)-1,3-dioxolan-4-yl]ethan-1-ol
SMILES CC(C)C1(OCC(CCO)O1)C(C)C

Specifications

Color Colorless
Quantity 1 g
Formula Weight 202.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.