accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH

CAS RN 1820711-54-3

IUPAC Name: N-(naphthalen-1-yl)-2,3-dihydro-1,4-benzodioxin-6-amine
Synonyms: N-(naphthalen-1-yl)-2,3-dihydro-1,4-benzodioxin-6-amine
Molecular Weight (g/mol): 277.32
Molecular Formula: C18H15NO2
InChi Key: WFXOBCJXPSQWNH-UHFFFAOYSA-N
SMILES: C1COC2=C(O1)C=CC(NC1=C3C=CC=CC3=CC=C1)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results