Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(1-Naphthylamino)-1,4-benzodioxane 97.0+%, TCI America™
SDP

Catalog No. N08331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08331G Supplier TCI America Supplier No. N08331G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1820711-54-3
Molecular Formula C18H15NO2
Molecular Weight (g/mol) 277.323
InChI Key WFXOBCJXPSQWNH-UHFFFAOYSA-N
Synonym N-[2,3-Dihydrobenzo[1,4]dioxin-6-yl]-1-naphthylamine
PubChem CID 44629971
IUPAC Name N-naphthalen-1-yl-2,3-dihydro-1,4-benzodioxin-6-amine
SMILES C1COC2=C(O1)C=CC(=C2)NC3=CC=CC4=CC=CC=C43

Specifications

Melting Point 135°C
Quantity 1 g
Formula Weight 277.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-(1-Naphthylamino)-1,4-benzodioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.