accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2N

CAS RN 1821-39-2

IUPAC Name: 2-propylaniline
Synonyms: 2-propyl-aniline
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: WKURVXXDGMYSDP-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC=C1N
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)