Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylaniline 98.0+%, TCI America™
SDP

Catalog No. P11135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11135G 5 g
P111325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11135G Supplier TCI America Supplier No. P11135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1821-39-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00009803
InChI Key WKURVXXDGMYSDP-UHFFFAOYSA-N
PubChem CID 15759
IUPAC Name 2-propylaniline
SMILES CCCC1=CC=CC=C1N

Specifications

Color Yellow
Boiling Point 223°C
Quantity 5 g
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Propylaniline
EINECSNumber (3)-0206
RTECSNumber BY8300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.