accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFFFFFFFF

CAS RN 1821062-80-9

IUPAC Name: 1,2,4,5-tetrafluoro-3-{3,3,4,4,5,5-hexafluoro-2-[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]cyclopent-1-en-1-yl}-6-(trifluoromethyl)benzene
Synonyms: 1,2,4,5-tetrafluoro-3-{3,3,4,4,5,5-hexafluoro-2-[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]cyclopent-1-en-1-yl}-6-(trifluoromethyl)benzene
Molecular Weight (g/mol): 608.18
Molecular Formula: C19F20
InChi Key: VDETZPYQAWUCIA-UHFFFAOYSA-N
SMILES: FC1=C(F)C(=C(F)C(F)=C1C1=C(C2=C(F)C(F)=C(C(F)=C2F)C(F)(F)F)C(F)(F)C(F)(F)C1(F)F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results