Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene 95.0+%, TCI America™
SDP

Catalog No. B5426500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5426500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5426500MG Supplier TCI America Supplier No. B5426500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1821062-80-9
Molecular Formula C19F20
Molecular Weight (g/mol) 608.177
InChI Key VDETZPYQAWUCIA-UHFFFAOYSA-N
Synonym 6,6′-(Perfluorocyclopent-1-ene-1,2-diyl)bis[1,2,4,5-tetrafluoro-3-(trifluoromethyl)benzene]
PubChem CID 122204810
IUPAC Name 1,2,4,5-tetrafluoro-3-[3,3,4,4,5,5-hexafluoro-2-[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]cyclopenten-1-yl]-6-(trifluoromethyl)benzene
SMILES C1(=C(C(=C(C(=C1F)F)C(F)(F)F)F)F)C2=C(C(C(C2(F)F)(F)F)(F)F)C3=C(C(=C(C(=C3F)F)C(F)(F)F)F)F

Specifications

Melting Point 72°C
Color White
Quantity 500 mg
Formula Weight 608.18
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.