accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18217-00-0

IUPAC Name: 1-(2-chloroethyl)-4-methoxybenzene
Synonyms: 1-(2-chloroethyl)-4-methoxybenzene
Molecular Weight (g/mol): 170.64
Molecular Formula: C9H11ClO
InChi Key: PMIAMRAWHYEPNH-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results