accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18227-41-3

IUPAC Name: 5-propylimidazolidine-2,4-dione
Synonyms: 5-propylimidazolidine-2,4-dione
Molecular Weight (g/mol): 142.16
Molecular Formula: C6H10N2O2
InChi Key: BIFASJFFCIDWDC-UHFFFAOYNA-N
SMILES: CCCC1NC(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results