Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Propylhydantoin 98.0+%, TCI America™
SDP

Catalog No. P032325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P032325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P032325G Supplier TCI America Supplier No. P032325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18227-41-3
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00059721
InChI Key BIFASJFFCIDWDC-UHFFFAOYNA-N
PubChem CID 567619
IUPAC Name 5-propylimidazolidine-2,4-dione
SMILES CCCC1NC(=O)NC1=O

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 25 g
Formula Weight 142.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Propylhydantoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.