accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ON

CAS RN 1822747-32-9

IUPAC Name: N-benzyl-3-methyl-N-phenylbenzamide
Synonyms: N-benzyl-3-methyl-N-phenylbenzamide
Molecular Weight (g/mol): 301.39
Molecular Formula: C21H19NO
InChi Key: LYTYTKCRBQDDKU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results