accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSBrFCl

CAS RN 1823304-13-7

IUPAC Name: methyl 7-bromo-3-chloro-4-fluoro-1-benzothiophene-2-carboxylate
Synonyms: methyl 7-bromo-3-chloro-4-fluoro-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 323.56
Molecular Formula: C10H5BrClFO2S
InChi Key: UHFJFXCPHLTILG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C2=C(F)C=CC(Br)=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results