accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 182344-29-2

IUPAC Name: {4'-ethoxy-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: 4'-ethoxy-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 242.08
Molecular Formula: C14H15BO3
InChi Key: ZFFXATWYKJBSCC-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results