accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHO

CAS RN 182491-21-0

IUPAC Name: 4-phenyloxane-4-carboxylic acid
Synonyms: 4-phenyloxane-4-carboxylic acid
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: BWHJLSRDMNLSET-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCOCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results