Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC69101DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC69101DA 1 g Amber Glass Bottle
CC69101CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC69101DA Supplier Thermo Scientific Chemicals Supplier No. CC69101DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 182491-21-0
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00085758
InChI Key BWHJLSRDMNLSET-UHFFFAOYSA-N
Synonym 4-phenyltetrahydro-2h-pyran-4-carboxylic acid, 4-phenyl-tetrahydro-pyran-4-carboxylic acid, 4-phenyltetrahydropyran-4-carboxylic acid, 4-phenyl-oxane-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-phenyl, 4-carboxy-4-phenyloxane, 4-carboxy-4-phenyltetrahydro-2h-pyran, 4-carboxytetrahydro-2h-pyran-4-yl benzene, 4-phenyltetrahydro-2h-pyran-4-carboxylicacid, 4-phenyl-tetrahydro-2h-pyran-4-carboxylic acid
PubChem CID 4138553
IUPAC Name 4-phenyloxane-4-carboxylic acid
SMILES OC(=O)C1(CCOCC1)C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 4-Phenyltetrahydropyran-4-carboxylic acid
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.