accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SHOSOHSH3CCH3CH3SOHCH3H3CCH3CH3CH3CH3OH

CAS RN 182496-55-5

IUPAC Name: 5,11,17,23-tetra-tert-butyl-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15(26),16,18,21,23-dodecaene-25,26,27,28-tetrol
Synonyms: 5,11,17,23-tetra-tert-butyl-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15(26),16,18,21,23-dodecaene-25,26,27,28-tetrol
Molecular Weight (g/mol): 721.06
Molecular Formula: C40H48O4S4
InChi Key: PDEJSTNRUYUEQL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(O)C(SC3=CC(=CC(SC4=C(O)C(SC5=C(O)C(S2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3O)C(C)(C)C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results