Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylthiacalix[4]arene 98.0+%, TCI America™
SDP

Catalog No. B22965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22965G 5 g
B22961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22965G Supplier TCI America Supplier No. B22965G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 182496-55-5
Molecular Formula C40H48O4S4
Molecular Weight (g/mol) 721.06
MDL Number MFCD04038412
InChI Key PDEJSTNRUYUEQL-UHFFFAOYSA-N
Synonym Tetra-tert-butyl(tetrahydroxy)tetrathiacalix[4]arene
PubChem CID 5242247
SMILES CC(C)(C)C1=CC2=C(C(=C1)SC3=CC(=CC(=C3O)SC4=CC(=CC(=C4O)SC5=C(C(=CC(=C5)C(C)(C)C)S2)O)C(C)(C)C)C(C)(C)C)O

Specifications

Melting Point 323°C
Color White-Yellow
Quantity 5 g
Formula Weight 721.06
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylthiacalix[4]arene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.