accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiCH3CH3H3C

CAS RN 1825-62-3

IUPAC Name: ethoxytrimethylsilane
Synonyms: ethoxytrimethylsilane
Molecular Weight (g/mol): 118.25
Molecular Formula: C5H14OSi
InChi Key: RSIHJDGMBDPTIM-UHFFFAOYSA-N
SMILES: CCO[Si](C)(C)C
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (5)