Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethoxytrimethylsilane

Catalog No. 254371100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-437-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-437-1100G Supplier Sigma Aldrich Supplier No. 254371100G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium acetylacetonate reacts with dichloro trimethylsiloxyaluminium to form ethoxytrimethylsilane.{70} Ethoxytrimethylsilane reacts with alkali ions to form Si-O-M (M: alkali metal) linkage, which was investigated in detail by 29Si NMR.1ETMS can be used to modify the silica nanoparticles to form a hydrophobic coating with ultra-high molecular weight polyethylene on the surface of the steel. It may be used to methylate silica nanoparticles that form silica nano-capsules for stabilized oil-in water emulsions.

Specifications

CAS 1825-62-3
Density 0.757 g/mL (at 25°C)
Boiling Point 75°C to 76°C
Molecular Formula C5H14OSi
Linear Formula C2H5OSi(CH3)3
Refractive Index n20/D 1.374 (literature)
MDL Number MFCD00009069
Quantity 100 g
Synonym Trimethylethoxysilane
Molecular Weight (g/mol) 118.25
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.