accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18251-82-6

IUPAC Name: bis(2-methylpropyl)amine hydrochloride
Synonyms: diisobutylamine hydrochloride
Molecular Weight (g/mol): 165.71
Molecular Formula: C8H20ClN
InChi Key: HDSBLUHKYYCLEZ-UHFFFAOYSA-N
SMILES: Cl.CC(C)CNCC(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results