Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D061525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D061525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D061525G Supplier TCI America Supplier No. D061525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18251-82-6
Molecular Formula C8H20ClN
Molecular Weight (g/mol) 165.705
MDL Number MFCD00060151
InChI Key HDSBLUHKYYCLEZ-UHFFFAOYSA-N
PubChem CID 519535
IUPAC Name 2-methyl-N-(2-methylpropyl)propan-1-amine;hydrochloride
SMILES CC(C)CNCC(C)C.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 165.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diisobutylamine Hydrochloride
EINECSNumber (2)-0137
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.