accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 182618-86-6

IUPAC Name: tert-butyl 4-(4-nitrophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(4-nitrophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 307.35
Molecular Formula: C15H21N3O4
InChi Key: MGYCIJUTYLUYJM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results