accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18282-40-1

IUPAC Name: 1-ethyl-2-iodobenzene
Synonyms: 1-ethyl-2-iodobenzene
Molecular Weight (g/mol): 232.06
Molecular Formula: C8H9I
InChi Key: ZEJZDNMOGNUIHL-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1I
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)