Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyliodobenzene 97.0+%, TCI America™
SDP

Catalog No. E05125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05125G Supplier TCI America Supplier No. E05125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18282-40-1
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00040871
InChI Key ZEJZDNMOGNUIHL-UHFFFAOYSA-N
Synonym 2-ethyliodobenzene, 1-iodo-2-ethylbenzene, benzene, 1-ethyl-2-iodo, 2-ethyl iodobenzene, pubchem3876, 1-ethyl-2-iodo-benzene, acmc-1brg2, #, benzene,1-ethyl-2-iodo, attercop-chm at134049
PubChem CID 140367
IUPAC Name 1-ethyl-2-iodobenzene
SMILES CCC1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 221°C
Quantity 5 g
Formula Weight 232.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyliodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.