accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClHO

CAS RN 1829-32-9

IUPAC Name: 3-chloro-2-hydroxybenzoic acid
Synonyms: 3-chloro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 172.56
Molecular Formula: C7H5ClO3
InChi Key: PPINMMULCRBDOS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(Cl)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results