accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COOBr

CAS RN 18291-80-0

IUPAC Name: trimethylsilyl 2-bromoacetate
Synonyms: trimethylsilyl 2-bromoacetate
Molecular Weight (g/mol): 211.13
Molecular Formula: C5H11BrO2Si
InChi Key: ZCHHFMWUDHXPFN-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results