Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl Bromoacetate 97.0+%, TCI America™
SDP

Catalog No. B157825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B157825G 25 g
B15785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B157825G Supplier TCI America Supplier No. B157825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18291-80-0
Molecular Formula C5H11BrO2Si
Molecular Weight (g/mol) 211.13
MDL Number MFCD00000187
InChI Key ZCHHFMWUDHXPFN-UHFFFAOYSA-N
Synonym trimethylsilyl bromoacetate, bromoacetic acid trimethylsilyl ester, acetic acid, bromo-, trimethylsilyl ester, acmc-209ek0, trimethylsilyl monobromoacetate, trimethylsilylbromoacetate, bromoacetic acid, tms derivative, zchhfmwudhxpfn-uhfffaoysa
PubChem CID 87554
IUPAC Name trimethylsilyl 2-bromoacetate
SMILES C[Si](C)(C)OC(=O)CBr

Specifications

Color Yellow
Boiling Point 83°C
Quantity 25 g
UN Number 2920
Formula Weight 211.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Bromoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.