accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3NNN

CAS RN 18293-54-4

IUPAC Name: 1-(trimethylsilyl)-1H-1,2,4-triazole
Synonyms: 1-(trimethylsilyl)-1,2,4-triazole
Molecular Weight (g/mol): 141.25
Molecular Formula: C5H11N3Si
InChi Key: WPSPBNRWECRRPK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N1C=NC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)