Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Trimethylsilyl-1,2,4-triazole, 95%

Catalog No. AAB2110514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2110514 25 g
AAB2110506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2110514 Supplier Thermo Scientific Chemicals Supplier No. B2110514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18293-54-4 | C5H11N3Si | 141.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18293-54-4
Molecular Formula C5H11N3Si
Molecular Weight (g/mol) 141.25
MDL Number MFCD00046070
InChI Key WPSPBNRWECRRPK-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1,2,4-triazole, 1-trimethylsilyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 1-trimethylsilyl, n-trimethylsilyl-1,2,4-triazole, acmc-20aorq, 1-trimethylsilyl-1,2,4 triazole, 1h-1,2,4-triazole, tms derivative, trimethyl 1,2,4-triazol-1-yl silane, 1h-1,2,4-triazole,1-trimethylsilyl
PubChem CID 87558
IUPAC Name trimethyl(1,2,4-triazol-1-yl)silane
SMILES C[Si](C)(C)N1C=NC=N1

Specifications

Density 0.98
Boiling Point 73°C to 75°C (12 mmHg)
Flash Point −5°C (23°F)
Refractive Index 1.461
Quantity 25 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 141.25
Percent Purity 98%
Chemical Name or Material 1-Trimethylsilyl-1,2,4-triazole
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 242-170-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.