accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18295-64-2

IUPAC Name: 10-chlorodec-3-yne
Synonyms: 10-chlorodec-3-yne
Molecular Weight (g/mol): 172.7
Molecular Formula: C10H17Cl
InChi Key: BQQIODGRFRSXTM-UHFFFAOYSA-N
SMILES: CCC#CCCCCCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results