Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Chloro-3-decyne 97.0+%, TCI America™
SDP

Catalog No. C15325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15325ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15325ML Supplier TCI America Supplier No. C15325ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18295-64-2
Molecular Formula C10H17Cl
Molecular Weight (g/mol) 172.696
MDL Number MFCD01321154
InChI Key BQQIODGRFRSXTM-UHFFFAOYSA-N
Synonym 10-chloro-3-decyne, 3-decyne, 10-chloro, chlorodecyne, 7-decyn-1-yl chloride, acmc-209ek5, bqqiodgrfrsxtm-uhfffaoysa
PubChem CID 87561
IUPAC Name 10-chlorodec-3-yne
SMILES CCC#CCCCCCCCl

Specifications

Color Colorless
Boiling Point 122°C
Quantity 5 mL
Formula Weight 172.70
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 10-Chloro-3-decyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.