accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 18299-85-9

IUPAC Name: docosyl prop-2-enoate
Synonyms: docosyl prop-2-enoate
Molecular Weight (g/mol): 380.66
Molecular Formula: C25H48O2
InChi Key: KHAYCTOSKLIHEP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCOC(=O)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results