Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Docosyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™
SDP

Catalog No. D5075100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5075100G 100 g
D507525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5075100G Supplier TCI America Supplier No. D5075100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18299-85-9
Molecular Formula C25H48O2
Molecular Weight (g/mol) 380.657
MDL Number MFCD00080409
InChI Key KHAYCTOSKLIHEP-UHFFFAOYSA-N
Synonym Acrylic Acid Docosyl Ester, Behenyl Acrylate, Acrylic Acid Behenyl Ester
PubChem CID 87565
IUPAC Name docosyl prop-2-enoate
SMILES CCCCCCCCCCCCCCCCCCCCCCOC(=O)C=C

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 100 g
Formula Weight 380.66
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Docosyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-3948
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.