accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 183065-68-1

IUPAC Name: 4-bromo-2,6-difluorobenzoic acid
Synonyms: 4-bromo-2,6-difluorobenzoic acid
Molecular Weight (g/mol): 237
Molecular Formula: C7H3BrF2O2
InChi Key: IRHPJGPQWZEZRX-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=C(Br)C=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results