Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-difluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B34475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34475G Supplier TCI America Supplier No. B34475G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 183065-68-1
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237.00
MDL Number MFCD03094085
InChI Key IRHPJGPQWZEZRX-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-bromobenzoic acid, benzoic acid, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzoic acid, 4-bromo-2,6-difluoro-benzoicacid, pubchem2476, acmc-1bx1y, ksc174k2f, rarechem al be 1306, benzoicacid, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluorobenzoic acid
PubChem CID 2773298
IUPAC Name 4-bromo-2,6-difluorobenzoic acid
SMILES OC(=O)C1=C(F)C=C(Br)C=C1F

Specifications

Melting Point 201°C
Quantity 5 g
Formula Weight 237.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,6-difluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.