accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 183322-16-9

IUPAC Name: ethyl 3,4-bis(2-methoxyethoxy)benzoate
Synonyms: ethyl 3,4-bis(2-methoxyethoxy)benzoate
Molecular Weight (g/mol): 298.34
Molecular Formula: C15H22O6
InChi Key: VGFZRAVMWXHEJB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(OCCOC)=C(OCCOC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results