accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3CH3

CAS RN 1838-73-9

IUPAC Name: 3-methylheptan-4-ol
Synonyms: 4-heptanol, 3-methyl-
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: JMRDKKYZLXDPLN-UHFFFAOYNA-N
SMILES: CCCC(O)C(C)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)