accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 183802-98-4

IUPAC Name: 5-bromo-2-chlorophenol
Synonyms: 5-bromo-2-chlorophenol
Molecular Weight (g/mol): 207.45
Molecular Formula: C6H4BrClO
InChi Key: UEVFFMZHGNYDKM-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)