accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18395-30-7

CH3OSiH3CCH3OCH3OH3C

CAS RN 18395-30-7

IUPAC Name: trimethoxy(2-methylpropyl)silane
Synonyms: trimethoxy(2-methylpropyl)silane
Molecular Weight (g/mol): 178.3
Molecular Formula: C7H18O3Si
InChi Key: XYJRNCYWTVGEEG-UHFFFAOYSA-N
SMILES: CO[Si](CC(C)C)(OC)OC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)